Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140734.1 | - | 14, 15 | 1511 | ||
AK220466.1 | - | 14, 15 | 1520 | |||
BC021396.1 | - | 13, 14 | 1550 | |||
BC070412.1 | - | 15, 16 | 1685 | 5705112 | ||
U96963.1 | - | 13, 14 | 1523 |