Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145779.1 | NP_001139251.1 | 9 | 894 | ||
NM_008442.2 | NP_032468.2 | 10 | 1277 | |||
GenBank mRNA | AK016720.1 | - | 9 | 897 | ||
AK037037.1 | - | 9 | 807 | |||
BC006803.1 | - | 9 | 889 | 3152883 | ||
D12644.1 | - | 10 | 1278 | |||
Y15894.1 | - | 8 | 744 |