Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010637.3 | NP_034767.2 | 3 | 1686 | ||
GenBank mRNA | AK141244.1 | - | 1 | 598 | ||
AK144942.1 | - | 3 | 1684 | |||
AK155343.1 | - | 3 | 1684 | |||
BC010301.1 | - | 3 | 1548 | 3156339 | ||
JF277566.1 | - | 2 | 1082 | |||
U20344.1 | - | 3 | 1392 | |||
U70662.1 | - | 3 | 1680 |