Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252658.1 | NP_001239587.1 | 3 | 410 | ||
NM_001252659.1 | NP_001239588.1 | 3 | 407 | |||
NM_010700.3 | NP_034830.2 | 3 | 410 | |||
GenBank mRNA | AF425607.1 | - | 3 | 258 | ||
AK018697.1 | - | 3 | 346 | |||
AK076243.1 | - | 3 | 388 | |||
AK149950.1 | - | 3 | 348 | |||
AK151968.1 | - | 3 | 377 | |||
AK152103.1 | - | 3 | 346 | |||
AK160006.1 | - | 3 | 348 | |||
AK161104.1 | - | 3 | 348 | |||
AK166466.1 | - | 3 | 347 | |||
AK170264.1 | - | 3 | 355 | |||
BC019207.1 | - | 3 | 328 | 5052599 | ||
BC053041.1 | - | 3 | 355 | 5704871 | ||
X64414.1 | - | 3 | 329 | |||
Z19521.1 | - | 3 | 208 |