Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168508.1 | NP_001161980.1 | 12 | 2993 | ||
NM_001168513.1 | NP_001161985.1 | 12 | 2594 | |||
NM_001168514.1 | NP_001161986.1 | 13 | 3038 | |||
NM_011951.3 | NP_036081.1 | 12 | 2993 | |||
GenBank mRNA | AK089059.1 | - | 12 | 2907 | ||
AK133684.1 | - | 12 | 2906 | |||
AK151348.1 | - | 12 | 2595 | |||
AK153025.1 | - | 12 | 2595 | |||
BC018227.1 | - | 13 | 2935 | |||
BC085303.1 | - | 12 | 2969 | 5389457 | ||
D83073.1 | - | 12 | 2823 | |||
U10871.1 | - | 12 | 2890 |