Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42ep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026514.2 | NP_080790.1 | 2 | 1444 | ||
GenBank mRNA | AK012309.1 | - | 2 | 1409 | ||
AK014281.1 | - | 2 | 1446 | |||
AK019447.1 | - | 2 | 1446 | |||
AK076782.1 | - | 2 | 1243 | |||
AK150214.1 | - | 2 | 1411 | |||
BC034714.1 | - | 2 | 1397 | 3595315 |