Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PADI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016233.2 | NP_057317.2 | 2 | 153 | ||
XM_011541571.2 | XP_011539873.1 | 2 | 1764 | |||
XM_011541572.2 | XP_011539874.1 | 2 | 172 | |||
GenBank mRNA | AB026831.1 | - | 2 | 154 | ||
BC041592.1 | - | 2 | 139 | 5431445 | ||
BC109091.1 | - | 2 | 140 | 40034659 | ||
BC109092.1 | - | 2 | 139 | 40034660 |