Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016237.4 | NP_057321.2 | 1, 2 | 337 | ||
XM_005253900.3 | XP_005253957.1 | 1, 2 | 339 | |||
XM_017019423.1 | XP_016874912.1 | 1, 2 | 55 | |||
XM_017019424.1 | XP_016874913.1 | 1, 2 | 55 | |||
GenBank mRNA | AF191339.1 | - | 1, 2 | 215 | ||
AI879332.1 | - | 1, 2 | 339 | |||
AK315451.1 | - | 1, 2 | 283 | |||
BC001081.2 | - | 1, 2 | 270 | 2823401 | ||
BC001950.1 | - | 1, 2 | 270 | 2823401 | ||
BC006301.2 | - | 1, 2 | 274 | 4100632 |