Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016245.4 | NP_057329.3 | 1 | 330 | ||
XM_011532021.1 | XP_011530323.1 | 1 | 279 | |||
GenBank mRNA | AF126780.1 | - | 1 | 285 | ||
AF273056.1 | - | 1 | 183 | |||
AK075348.1 | - | 1 | 214 | |||
AY358553.1 | - | 1 | 197 | |||
BC008650.1 | - | 1 | 220 | 3854750 | ||
BC014327.1 | - | 1 | 196 | 3961721 | ||
BC016367.1 | - | 1 | 227 | 4133318 | ||
BC021673.1 | - | 1 | 201 | 3827935 | ||
BC036001.1 | - | 1 | 221 | 4715042 | ||
BU838518.1 | - | 412 | ||||
DA265697.1 | - | 1 | 329 |