Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG2 SIRNA Assays ›
Gene Symbol:
NDRG2
Gene Name:
NDRG family member 2
Gene Aliases:
SYLD
Chromosome Location:
Chr.14: 21016763 - 21070872 on Build GRCh38
UniGene ID:
Hs.525205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24073
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282211.1
NP_001269140.1
4
275
NM_001282212.1
NP_001269141.1
5
549
NM_001282213.1
NP_001269142.1
5
554
NM_001282214.1
NP_001269143.1
5
542
NM_001282215.1
NP_001269144.1
5
372
NM_001282216.1
NP_001269145.1
4
290
NM_001320329.1
NP_001307258.1
5
1207
NM_016250.2
NP_057334.1
5
397
NM_201535.1
NP_963293.1
6
439
NM_201536.1
NP_963294.1
4
330
NM_201537.1
NP_963831.1
5
372
NM_201538.1
NP_963832.1
4
307
NM_201539.1
NP_963833.1
5
349
NM_201540.1
NP_963834.1
6
416
NM_201541.1
NP_963835.1
5
374
XM_011536996.2
XP_011535298.1
6
426
XM_011536997.1
XP_011535299.1
6
427
XM_011536998.1
XP_011535300.1
6
417
XM_011536999.1
XP_011535301.1
4
822
XM_011537001.1
XP_011535303.1
5
380
XM_011537002.1
XP_011535304.1
5
526
XM_017021480.1
XP_016876969.1
6
378
XM_017021481.1
XP_016876970.1
6
417
XM_017021482.1
XP_016876971.1
6
426
XM_017021483.1
XP_016876972.1
5
310
XM_017021484.1
XP_016876973.1
5
352
XM_017021485.1
XP_016876974.1
6
417
XM_017021486.1
XP_016876975.1
6
427
XM_017021487.1
XP_016876976.1
5
336
XM_017021488.1
XP_016876977.1
5
377
XM_017021489.1
XP_016876978.1
5
482
XM_017021490.1
XP_016876979.1
4
274
XM_017021491.1
XP_016876980.1
4
309
XM_017021492.1
XP_016876981.1
5
380
XM_017021493.1
XP_016876982.1
5
526
XM_017021494.1
XP_016876983.1
6
378
XM_017021495.1
XP_016876984.1
5
310
XM_017021496.1
XP_016876985.1
4
309
GenBank mRNA
AB033074.1
-
4
259
AF087872.1
-
4
313
AF159092.3
-
5
320
AF304051.1
-
6
439
AK024521.1
-
6
372
AK057843.1
-
5
343
AK057847.1
-
4
307
AK090657.1
-
4
246
AK096999.1
-
5
1207
AK130029.1
-
4
245
AK293514.1
-
4
275
AL136574.1
-
4
284
BC010458.1
-
4
278
4215141
BC011240.1
-
5
332
4153602
BC093038.1
-
5
373
4811718
BG818977.1
-
5
353
BI759006.1
-
6
358
BM810623.1
-
5
313
BX247987.1
-
5
408
BX248031.1
-
3
236
BX647091.1
-
2
472
BX647748.1
-
5
489
CB153966.1
-
4
373
CR749252.1
-
5
542
DA065447.1
-
5
549
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbzcs:80/10.239.166.68:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline