Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016256.3 | NP_057340.2 | 3, 4 | 710 | ||
XM_011522517.2 | XP_011520819.1 | 3, 4 | 710 | |||
GenBank mRNA | AF187072.1 | - | 3, 4 | 683 | ||
AK025570.1 | - | 3, 4 | 709 | |||
AK026122.1 | - | 3, 4 | 686 | |||
AK127952.1 | - | 3, 4 | 704 | |||
AK314320.1 | - | 3, 4 | 704 | |||
BC012194.1 | - | 3, 4 | 700 | 4661335 |