Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006956.2 | NP_008887.2 | 4 | 773 | ||
NM_016265.3 | NP_057349.2 | 4 | 773 | |||
GenBank mRNA | AB021643.1 | - | 4 | 773 | ||
AF505656.1 | - | 4 | 440 | |||
AK293925.1 | - | 5 | 419 | |||
AY500354.1 | - | 1 | 75 | |||
BC051892.1 | - | 4 | 761 | |||
BC137068.1 | - | 3 | 265 | 9021065 | ||
BC137069.1 | - | 3 | 265 | 9021066 | ||
BC144465.1 | - | 3 | 265 | |||
BC154414.1 | - | 4 | 761 | 40148925 | ||
BX640749.1 | - | 3 | 298 |