Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016305.3 | NP_057389.1 | 3 | 294 | ||
XM_017006548.1 | XP_016862037.1 | 2 | 1306 | |||
XM_017006549.1 | XP_016862038.1 | 3 | 265 | |||
GenBank mRNA | AF201950.1 | - | 2 | 184 | ||
AK312225.1 | - | 2 | 89 | |||
BC017804.1 | - | 2 | 112 | 4658643 | ||
BC020444.1 | - | 2 | 84 | |||
BX114027.1 | - | 3 | 288 | |||
EL954786.1 | - | 3 | 297 |