Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016306.5 | NP_057390.1 | 2 | 359 | ||
GenBank mRNA | AB028859.2 | - | 2 | 302 | ||
AF228505.1 | - | 2 | 228 | |||
AF277317.1 | - | 2 | 319 | |||
AJ250137.1 | - | 2 | 157 | |||
AK075300.1 | - | 2 | 316 | |||
AK075430.1 | - | 2 | 335 | |||
AY359043.1 | - | 2 | 295 | |||
BC001144.1 | - | 2 | 297 | 3534756 | ||
BC046500.1 | - | 3 | 1066 | |||
BM754506.1 | - | 2 | 346 | |||
BT007063.1 | - | 2 | 137 | |||
CR457096.1 | - | 2 | 137 | |||
DA058195.1 | - | 2 | 359 |