Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303533.1 | NP_001290462.1 | 3, 4 | 336 | ||
NM_001303534.1 | NP_001290463.1 | 3, 4 | 431 | |||
NM_016339.4 | NP_057423.1 | 3, 4 | 616 | |||
XM_006721932.3 | XP_006721995.1 | 3, 4 | 1110 | |||
XM_006721935.3 | XP_006721998.1 | 4, 5 | 387 | |||
XM_006721937.3 | XP_006722000.1 | 3, 4 | 2212 | |||
GenBank mRNA | AF117946.1 | - | 3, 4 | 251 | ||
AK304160.1 | - | 3, 4 | 431 | |||
AK313492.1 | - | 3, 4 | 291 | |||
BC029956.1 | - | 3, 4 | 1110 | |||
BC115374.1 | - | 1, 2 | 145 | 40028278 | ||
BC115375.1 | - | 1, 2 | 145 | 40028280 | ||
DB265550.1 | - | 3, 4 | 338 |