Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016347.2 | NP_057431.2 | 1 | 660 | ||
NR_132338.1 | 1 | 660 | ||||
GenBank mRNA | AF185571.1 | - | 1 | 660 | ||
BC069564.1 | - | 1 | 660 | |||
BC121101.1 | - | 1 | 664 | |||
BC121102.1 | - | 1 | 664 | |||
BX105650.1 | - | 1 | 140 |