Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF280D SIRNA Assays ›
Gene Symbol:
ZNF280D
Gene Name:
zinc finger protein 280D
Gene Aliases:
SUHW4, ZNF634
Chromosome Location:
Chr.15: 56630176 - 56734086 on Build GRCh38
UniGene ID:
Hs.511477
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241351
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF280D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002843.2
NP_001002843.1
5
966
NM_001002844.2
NP_001002844.1
6
1028
NM_001288588.1
NP_001275517.1
6
1028
NM_001288589.1
NP_001275518.1
6
1035
NM_017661.3
NP_060131.2
6
1035
XM_005254481.3
XP_005254538.2
5
458
XM_005254483.3
XP_005254540.2
5
458
XM_005254484.3
XP_005254541.2
5
458
XM_005254485.3
XP_005254542.2
5
458
XM_011521701.1
XP_011520003.1
5
458
XM_011521702.1
XP_011520004.1
7
508
XM_011521704.2
XP_011520006.1
5
458
XM_011521707.2
XP_011520009.1
5
458
XM_011521711.1
XP_011520013.1
5
458
XM_017022344.1
XP_016877833.1
6
550
XM_017022345.1
XP_016877834.1
6
946
XM_017022346.1
XP_016877835.1
5
458
XM_017022347.1
XP_016877836.1
6
543
XM_017022348.1
XP_016877837.1
6
511
GenBank mRNA
AB046804.1
-
5
370
AK289739.1
-
5
389
AK295159.1
-
5
510
AK310882.1
-
6
456
BC015382.2
-
4
362
BC040532.1
-
5
967
BC053649.1
-
6
434
BC064359.1
-
6
436
BC130451.1
-
6
498
40146482
BC136412.1
-
6
491
9020400
BX640637.1
-
6
483
BX640707.1
-
6
539
BX641000.1
-
5
460
CX788386.1
-
6
436
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
axon guidance
negative regulation of transcription, DNA-templated
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0