Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001247997.1 | NP_001234926.1 | 8 | 1507 | ||
NM_002956.2 | NP_002947.1 | 8 | 1507 | |||
NM_198240.1 | NP_937883.1 | 8 | 1507 | |||
XM_005253593.2 | XP_005253650.1 | 8 | 1570 | |||
XM_006719552.3 | XP_006719615.1 | 8 | 1596 | |||
XM_017019786.1 | XP_016875275.1 | 8 | 1671 | |||
XM_017019787.1 | XP_016875276.1 | 8 | 1570 | |||
XM_017019788.1 | XP_016875277.1 | 8 | 1596 | |||
XM_017019789.1 | XP_016875278.1 | 8 | 1755 | |||
XM_017019790.1 | XP_016875279.1 | 8 | 1497 | |||
XM_017019791.1 | XP_016875280.1 | 8 | 1596 | |||
XM_017019792.1 | XP_016875281.1 | 8 | 1596 | |||
GenBank mRNA | AB209115.1 | - | 5 | 605 | ||
AF045652.1 | - | 2 | 62 | |||
AF045653.1 | - | 2 | 62 | |||
AY563000.1 | - | 2 | 77 | |||
BC047895.1 | - | 7 | 1315 | |||
BC062543.1 | - | 7 | 1463 | |||
BC114213.1 | - | 7 | 1422 | 7961952 | ||
BC117209.1 | - | 7 | 1394 | 40125760 | ||
BC126305.1 | - | 7 | 1394 | 8992021 | ||
M97501.1 | - | 8 | 1507 | |||
X64838.1 | - | 8 | 1484 |