Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016215.4 | NP_057299.1 | 9 | 1133 | ||
NM_201446.2 | NP_958854.1 | 7 | 687 | |||
NR_045110.1 | 8 | 914 | ||||
NR_045111.1 | 8 | 917 | ||||
NR_046367.1 | 8 | 1510 | ||||
XM_006717141.3 | XP_006717204.1 | 6 | 514 | |||
XM_011518764.1 | XP_011517066.1 | 9 | 1834 | |||
XM_011518765.1 | XP_011517067.1 | 9 | 1831 | |||
XM_011518766.1 | XP_011517068.1 | 10 | 2050 | |||
XM_011518767.1 | XP_011517069.1 | 6 | 1272 | |||
XM_011518768.1 | XP_011517070.1 | 6 | 1269 | |||
XM_017014795.1 | XP_016870284.1 | 7 | 1596 | |||
XM_017014796.1 | XP_016870285.1 | 7 | 1593 | |||
XM_017014797.1 | XP_016870286.1 | 8 | 1672 | |||
XM_017014798.1 | XP_016870287.1 | 8 | 1809 | |||
XM_017014799.1 | XP_016870288.1 | 8 | 1812 | |||
GenBank mRNA | AB125649.1 | - | 7 | 669 | ||
AF186111.1 | - | 6 | 618 | |||
AK091964.1 | - | 8 | 1510 | |||
AL512735.1 | - | 8 | 914 | |||
AY358902.1 | - | 9 | 1086 | |||
AY358903.1 | - | 8 | 924 | |||
BC012377.1 | - | 7 | 667 | 3860522 | ||
BC088371.1 | - | 8 | 907 | 30400137 | ||
BM919940.1 | - | 7 | 687 | |||
BX647866.1 | - | 1 | 301 | |||
KC485578.1 | - | 6 | 599 |