Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256012.1 | NP_001242941.1 | 9 | 1003 | ||
NM_001256095.1 | NP_001243024.1 | 9 | 1000 | |||
NM_005964.3 | NP_005955.3 | 8 | 973 | |||
XM_011523875.2 | XP_011522177.1 | 9 | 1292 | |||
XM_011523878.2 | XP_011522180.1 | 9 | 1344 | |||
XM_011523879.2 | XP_011522181.1 | 9 | 1289 | |||
XM_011523880.2 | XP_011522182.1 | 8 | 1262 | |||
XM_017024677.1 | XP_016880166.1 | 9 | 1058 | |||
XM_017024678.1 | XP_016880167.1 | 10 | 1040 | |||
XM_017024679.1 | XP_016880168.1 | 10 | 1081 | |||
XM_017024680.1 | XP_016880169.1 | 10 | 1389 | |||
XM_017024681.1 | XP_016880170.1 | 9 | 1010 | |||
XM_017024682.1 | XP_016880171.1 | 9 | 985 | |||
GenBank mRNA | AB210026.1 | - | 9 | 955 | ||
BC117690.1 | - | 7 | 875 | 40072059 | ||
BC117691.1 | - | 7 | 875 | 40072061 | ||
BC144668.1 | - | 9 | 957 | 9053199 | ||
BC150634.1 | - | 9 | 957 | 9057004 | ||
BG421687.1 | - | 4 | 244 | |||
M69181.1 | - | 8 | 913 | |||
U15618.1 | - | 5 | 249 |