Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135643.1 | NP_001129115.1 | 4 | 512 | ||
NM_001135644.1 | NP_001129116.1 | 4 | 680 | |||
NM_016221.3 | NP_057305.1 | 4 | 512 | |||
XM_011537644.1 | XP_011535946.1 | 5 | 416 | |||
XM_011537645.1 | XP_011535947.1 | 5 | 328 | |||
XM_011537646.1 | XP_011535948.1 | 5 | 549 | |||
GenBank mRNA | AF195120.1 | - | 4 | 427 | ||
AK000299.1 | - | 4 | 431 | |||
AK125973.1 | - | 4 | 680 | |||
AK299517.1 | - | 4 | 555 | |||
AK307757.1 | - | 4 | 451 | |||
BC026323.1 | - | 4 | 437 | 4815273 | ||
DC321382.1 | - | 4 | 512 | |||
KU178637.1 | - | 4 | 410 | |||
KU178638.1 | - | 4 | 410 |