Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251987.1 | NP_001238916.1 | 5 | 775 | ||
NM_001256020.1 | NP_001242949.1 | 5 | 775 | |||
NM_001256021.1 | NP_001242950.1 | 5 | 775 | |||
NM_001256022.1 | NP_001242951.1 | 5 | 775 | |||
NM_006073.3 | NP_006064.2 | 5 | 775 | |||
GenBank mRNA | AJ489257.2 | - | 5 | 670 | ||
AK291867.1 | - | 5 | 632 | |||
AK307619.1 | - | 5 | 452 | |||
BC029393.1 | - | 5 | 632 | |||
BC057222.1 | - | 5 | 636 | |||
BC070290.1 | - | 5 | 654 | |||
BC139910.1 | - | 5 | 591 | |||
JN900469.1 | - | 5 | 589 | |||
U18985.1 | - | 5 | 479 |