Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016224.4 | NP_057308.1 | 3 | 384 | ||
XM_005267015.2 | XP_005267072.1 | 5 | 462 | |||
GenBank mRNA | AF121859.1 | - | 3 | 144 | ||
AF131214.1 | - | 3 | 163 | |||
AF172847.1 | - | 3 | 264 | |||
BC005022.1 | - | 3 | 291 | 3832234 | ||
BG766499.1 | - | 3 | 294 | |||
DA315945.1 | - | 3 | 385 | |||
DA336311.1 | - | 3 | 330 |