Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007137.3 | NP_009068.2 | 5 | 542 | ||
XM_005272600.3 | XP_005272657.1 | 5 | 600 | |||
XM_011543899.2 | XP_011542201.1 | 5 | 2206 | |||
XM_011543900.2 | XP_011542202.1 | 6 | 661 | |||
XM_017029486.1 | XP_016884975.1 | 6 | 719 | |||
XM_017029487.1 | XP_016884976.1 | 5 | 952 | |||
GenBank mRNA | AK126949.1 | - | 6 | 661 | ||
AY487248.1 | - | 4 | 293 | |||
DR002896.1 | - | 5 | 530 |