Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199933.1 | NP_001186862.1 | 10 | 2525 | ||
NM_001199934.1 | NP_001186863.1 | 10 | 2567 | |||
NM_014454.2 | NP_055269.1 | 10 | 3039 | |||
GenBank mRNA | AF033120.1 | - | 10 | 2518 | ||
AF033121.1 | - | 10 | 2567 | |||
AF033122.1 | - | 10 | 2618 | |||
AJ420599.1 | - | 1 | 1041 | |||
AK001886.1 | - | 5 | 1521 | |||
AL133559.1 | - | 1 | 833 |