Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199933.1 | NP_001186862.1 | 10 | 1566 | ||
NM_001199934.1 | NP_001186863.1 | 10 | 1608 | |||
NM_014454.2 | NP_055269.1 | 10 | 2080 | |||
GenBank mRNA | AF033120.1 | - | 10 | 1559 | ||
AF033121.1 | - | 10 | 1608 | |||
AF033122.1 | - | 10 | 1659 | |||
AJ420599.1 | - | 1 | 82 | |||
AK001886.1 | - | 5 | 562 | |||
AK290157.1 | - | 10 | 1887 | |||
AK290551.1 | - | 10 | 1563 | |||
BC112036.1 | - | 10 | 1712 | 8327504 | ||
BC113569.1 | - | 10 | 1700 | 8322621 |