Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077440.1 | NP_001070908.1 | 6 | 2001 | ||
NM_001077441.1 | NP_001070909.1 | 6 | 1488 | |||
NM_001301038.1 | NP_001287967.1 | 6 | 2001 | |||
NM_014739.2 | NP_055554.1 | 6 | 2007 | |||
XM_005267237.3 | XP_005267294.1 | 6 | 1943 | |||
XM_017011529.1 | XP_016867018.1 | 6 | 1475 | |||
XM_017011530.1 | XP_016867019.1 | 6 | 2282 | |||
XM_017011531.1 | XP_016867020.1 | 6 | 1418 | |||
XM_017011532.1 | XP_016867021.1 | 6 | 1943 | |||
GenBank mRNA | AF249273.1 | - | 4 | 1748 | ||
BC016682.1 | - | 6 | 1946 | |||
BC047687.1 | - | 5 | 1314 | |||
BC047887.1 | - | 6 | 1976 | |||
BC056894.1 | - | 6 | 1988 | |||
BC063846.1 | - | 6 | 1914 | |||
BC078160.1 | - | 6 | 1913 | |||
BC132780.1 | - | 6 | 1951 | 40146802 | ||
BC144281.1 | - | 6 | 1945 | 9052801 | ||
BC144299.1 | - | 6 | 1951 | |||
BX648878.1 | - | 2 | 748 | |||
BX649160.1 | - | 6 | 1419 | |||
D79986.1 | - | 6 | 2007 |