Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142864.3 | NP_001136336.2 | 51 | 7661 | ||
GenBank mRNA | AB161230.1 | - | 40 | 6112 | ||
AK092007.1 | - | 4 | 2114 | |||
BC008073.2 | - | 10 | 1212 | |||
BC150271.1 | - | 40 | 5949 | 9007243 | ||
D87071.1 | - | 40 | 5949 | |||
KC602455.1 | - | 51 | 7402 |