Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164814.1 | NP_001158286.1 | 19 | 4429 | ||
NM_001164815.1 | NP_001158287.1 | 18 | 4348 | |||
NM_001164816.1 | NP_001158288.1 | 12 | 1987 | |||
NM_001164817.1 | NP_001158289.1 | 13 | 2049 | |||
NM_014977.3 | NP_055792.1 | 19 | 4468 | |||
XM_005267415.3 | XP_005267472.1 | 19 | 4269 | |||
XM_005267416.3 | XP_005267473.1 | 19 | 4236 | |||
XM_005267418.1 | XP_005267475.1 | 12 | 2284 | |||
GenBank mRNA | AB014570.2 | - | 19 | 3977 | ||
AF124726.1 | - | 19 | 4467 | |||
AF124727.1 | - | 13 | 2003 | |||
AF124728.1 | - | 12 | 1978 | |||
AL050382.1 | - | 12 | 3073 | |||
AL833228.1 | - | 14 | 2318 | |||
BX247975.1 | - | 12 | 2257 |