Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016234.3 | NP_057318.2 | 4, 5 | 618 | ||
NM_203379.1 | NP_976313.1 | 4, 5 | 479 | |||
NM_203380.1 | NP_976314.1 | 4, 5 | 645 | |||
XM_017016324.1 | XP_016871813.1 | 4, 5 | 653 | |||
GenBank mRNA | AB033899.1 | - | 4, 5 | 492 | ||
AK000339.1 | - | 4, 5 | 628 | |||
AK024932.1 | - | 4, 5 | 619 | |||
AK222782.1 | - | 4, 5 | 636 | |||
AK223288.1 | - | 4, 5 | 607 | |||
AK310436.1 | - | 4, 5 | 428 | |||
AM262166.1 | - | 3, 4 | 338 | |||
AY358520.1 | - | 4, 5 | 579 | |||
BC007985.2 | - | 4, 5 | 596 | 2959548 | ||
CF595490.1 | - | 4, 5 | 482 |