Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015210.3 | NP_056025.2 | 9 | 2238 | ||
XM_005258098.3 | XP_005258155.1 | 7 | 3138 | |||
XM_005258099.4 | XP_005258156.1 | 7 | 3138 | |||
XM_011525640.2 | XP_011523942.1 | 9 | 3339 | |||
XM_011525641.2 | XP_011523943.1 | 8 | 3261 | |||
XM_017025671.1 | XP_016881160.1 | 9 | 2708 | |||
XM_017025672.1 | XP_016881161.1 | 9 | 2309 | |||
XM_017025673.1 | XP_016881162.1 | 10 | 2316 | |||
XM_017025674.1 | XP_016881163.1 | 9 | 2363 | |||
XM_017025675.1 | XP_016881164.1 | 9 | 2238 | |||
GenBank mRNA | AB018345.2 | - | 7 | 2155 | ||
AK131528.1 | - | 9 | 2238 | |||
BC040542.1 | - | 2 | 135 | 5268201 |