Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 5 | 583 | ||
XM_005268750.4 | XP_005268807.1 | 5 | 423 | |||
XM_011538083.1 | XP_011536385.1 | 5 | 447 | |||
XM_017019091.1 | XP_016874580.1 | 5 | 423 | |||
XM_017019092.1 | XP_016874581.1 | 4 | 327 | |||
XM_017019093.1 | XP_016874582.1 | 4 | 276 | |||
XM_017019094.1 | XP_016874583.1 | 4 | 304 | |||
GenBank mRNA | AB023163.1 | - | 5 | 527 | ||
AB024494.1 | - | 5 | 1070 | |||
AB097013.1 | - | 5 | 403 | |||
AB097035.1 | - | 5 | 403 | |||
AF049612.1 | - | 5 | 416 | |||
AF161412.1 | - | 5 | 415 | |||
AK292866.1 | - | 5 | 399 | |||
AK299089.1 | - | 4 | 295 | |||
BC030990.1 | - | 4 | 450 | |||
BC050324.1 | - | 5 | 542 | 5266097 |