Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 7 | 868 | ||
XM_005268750.4 | XP_005268807.1 | 7 | 667 | |||
XM_011538083.1 | XP_011536385.1 | 7 | 732 | |||
XM_017019091.1 | XP_016874580.1 | 7 | 708 | |||
XM_017019092.1 | XP_016874581.1 | 6 | 571 | |||
XM_017019093.1 | XP_016874582.1 | 6 | 520 | |||
XM_017019094.1 | XP_016874583.1 | 6 | 548 | |||
XM_017019095.1 | XP_016874584.1 | 3 | 260 | |||
GenBank mRNA | AB023163.1 | - | 7 | 812 | ||
AB024494.1 | - | 7 | 1358 | |||
AB097013.1 | - | 7 | 688 | |||
AB097035.1 | - | 7 | 688 | |||
AK292866.1 | - | 7 | 684 | |||
AK299089.1 | - | 6 | 580 | |||
BC030990.1 | - | 6 | 735 | |||
BC050324.1 | - | 7 | 827 | 5266097 |