Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 30 | 6019 | ||
XM_005267502.2 | XP_005267559.2 | 30 | 6181 | |||
XM_011536619.2 | XP_011534921.1 | 30 | 5611 | |||
XM_011536621.2 | XP_011534923.1 | 29 | 4780 | |||
XM_011536622.1 | XP_011534924.1 | 21 | 3944 | |||
XM_017021148.1 | XP_016876637.1 | 29 | 5370 | |||
XM_017021149.1 | XP_016876638.1 | 29 | 5364 | |||
XM_017021150.1 | XP_016876639.1 | 29 | 5511 | |||
XM_017021151.1 | XP_016876640.1 | 29 | 5376 | |||
XM_017021152.1 | XP_016876641.1 | 29 | 5370 | |||
XM_017021153.1 | XP_016876642.1 | 29 | 4774 | |||
GenBank mRNA | AB032957.2 | - | 22 | 4165 | ||
AK024847.1 | - | 8 | 1848 | |||
AK095634.1 | - | 21 | 3771 | |||
AK125788.1 | - | 4 | 1126 | |||
AL110222.1 | - | 5 | 537 | |||
AY254380.1 | - | 30 | 5819 | |||
AY739714.1 | - | 6 | 1333 |