Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 42 | 8162 | ||
XM_005267502.2 | XP_005267559.2 | 42 | 8324 | |||
XM_011536619.2 | XP_011534921.1 | 42 | 7754 | |||
XM_011536621.2 | XP_011534923.1 | 41 | 6923 | |||
XM_011536622.1 | XP_011534924.1 | 33 | 6087 | |||
XM_017021148.1 | XP_016876637.1 | 41 | 7513 | |||
XM_017021149.1 | XP_016876638.1 | 41 | 7507 | |||
XM_017021150.1 | XP_016876639.1 | 40 | 7471 | |||
XM_017021151.1 | XP_016876640.1 | 41 | 7519 | |||
XM_017021152.1 | XP_016876641.1 | 41 | 7513 | |||
XM_017021153.1 | XP_016876642.1 | 41 | 6917 | |||
GenBank mRNA | AB032957.2 | - | 34 | 6308 | ||
AK024847.1 | - | 20 | 3991 | |||
AK125788.1 | - | 16 | 3269 | |||
AL110222.1 | - | 17 | 2680 | |||
AY254380.1 | - | 42 | 7962 | |||
AY739714.1 | - | 18 | 3476 | |||
BC006237.2 | - | 1 | 167 | |||
BC011658.2 | - | 7 | 1363 | |||
BC063686.1 | - | 5 | 845 | |||
BC072441.1 | - | 5 | 916 | |||
BC083494.1 | - | 3 | 317 |