Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204197.1 | NP_001191126.1 | 8 | 937 | ||
NM_015678.4 | NP_056493.3 | 48 | 7967 | |||
XM_005266346.4 | XP_005266403.1 | 48 | 7967 | |||
XM_005266347.4 | XP_005266404.2 | 48 | 7967 | |||
XM_005266348.4 | XP_005266405.2 | 47 | 7958 | |||
XM_006719803.3 | XP_006719866.1 | 48 | 7967 | |||
XM_006719805.3 | XP_006719868.1 | 47 | 7958 | |||
XM_006719806.3 | XP_006719869.1 | 47 | 7958 | |||
XM_011535046.2 | XP_011533348.1 | 40 | 6098 | |||
XM_017020544.1 | XP_016876033.1 | 48 | 7949 | |||
XM_017020545.1 | XP_016876034.1 | 48 | 7949 | |||
XM_017020546.1 | XP_016876035.1 | 48 | 7255 | |||
GenBank mRNA | AB046764.1 | - | 15 | 1679 | ||
AF072371.1 | - | 2 | 154 | |||
AF467288.1 | - | 49 | 7639 | |||
AK091469.1 | - | 19 | 2126 | |||
AK122888.1 | - | 18 | 2480 | |||
AK127791.1 | - | 25 | 3318 | |||
AK294737.1 | - | 8 | 937 | |||
AK297773.1 | - | 9 | 1035 | |||
AK315939.1 | - | 8 | 934 | |||
AL137748.1 | - | 2 | 579 | |||
BX640760.1 | - | 24 | 3297 |