Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136197.1 | NP_001129669.1 | 10 | 1196 | ||
NM_001136198.1 | NP_001129670.1 | 12 | 1463 | |||
NM_016263.3 | NP_057347.2 | 13 | 1641 | |||
XM_005259573.4 | XP_005259630.1 | 13 | 1823 | |||
XM_017026863.1 | XP_016882352.1 | 14 | 2167 | |||
GenBank mRNA | AB013462.1 | - | 12 | 1429 | ||
AB013463.1 | - | 10 | 1162 | |||
AB033068.1 | - | 12 | 1463 | |||
AF080397.1 | - | 12 | 1441 | |||
AF083810.1 | - | 13 | 1568 | |||
AF102508.1 | - | 3 | 378 | |||
AF433157.1 | - | 10 | 1162 | |||
BC013413.2 | - | 13 | 1618 | 3160334 | ||
BT007115.1 | - | 12 | 1429 |