Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002897.4 | NP_002888.1 | 5 | 869 | ||
NM_016836.3 | NP_058520.1 | 5 | 869 | |||
XM_005246737.1 | XP_005246794.1 | 5 | 580 | |||
XM_005246738.2 | XP_005246795.1 | 5 | 915 | |||
XM_005246739.2 | XP_005246796.1 | 5 | 785 | |||
XM_005246740.1 | XP_005246797.1 | 5 | 580 | |||
XM_005246741.3 | XP_005246798.1 | 5 | 782 | |||
XM_017004626.1 | XP_016860115.1 | 5 | 915 | |||
GenBank mRNA | AK225887.1 | - | 5 | 480 | ||
AK297812.1 | - | 5 | 580 | |||
AK309949.1 | - | 5 | 544 | |||
BC012992.2 | - | 5 | 564 | 3530897 | ||
BC012993.2 | - | 5 | 564 | 3530898 | ||
BC018951.2 | - | 5 | 856 | 4137722 | ||
BC051889.1 | - | 5 | 451 | |||
D28482.1 | - | 5 | 435 | |||
KU178305.1 | - | 5 | 425 | |||
KU178306.1 | - | 5 | 422 | |||
L11289.1 | - | 5 | 493 | |||
X64652.1 | - | 4 | 378 | |||
X77494.1 | - | 6 | 557 |