Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286721.1 | NP_001273650.1 | 2 | 397 | ||
NM_017664.3 | NP_060134.2 | 2 | 397 | |||
NR_104586.1 | 2 | 397 | ||||
NR_104587.1 | 2 | 397 | ||||
GenBank mRNA | AK000100.1 | - | 2 | 359 | ||
AK096083.1 | - | 2 | 317 | |||
BC001727.1 | - | 2 | 296 | 3354202 | ||
BC039715.1 | - | 2 | 327 | |||
BX354283.2 | - | 2 | 293 | |||
DA961131.1 | - | 2 | 397 |