Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286721.1 | NP_001273650.1 | 2, 3 | 547 | ||
NM_017664.3 | NP_060134.2 | 2, 3 | 547 | |||
NR_104586.1 | 2, 3 | 547 | ||||
NR_104587.1 | 2, 3 | 547 | ||||
GenBank mRNA | AK000100.1 | - | 2, 3 | 509 | ||
BC001727.1 | - | 2, 3 | 446 | 3354202 | ||
BC039715.1 | - | 2, 3 | 477 | |||
BX354283.2 | - | 2, 3 | 443 |