Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146189.1 | NP_001139661.1 | 1 | 4100 | ||
NM_001146190.1 | NP_001139662.1 | 1 | 4100 | |||
NM_017757.2 | NP_060227.2 | 1 | 4100 | |||
XM_005266726.4 | XP_005266783.1 | 2 | 4299 | |||
XM_006722500.3 | XP_006722563.1 | 2 | 4164 | |||
XM_011526068.2 | XP_011524370.1 | 2 | 7806 | |||
XM_011526069.2 | XP_011524371.1 | 2 | 4164 | |||
XM_011526070.1 | XP_011524372.1 | 2 | 4164 | |||
XM_017025838.1 | XP_016881327.1 | 2 | 4164 | |||
GenBank mRNA | AB051490.1 | - | 1 | 796 | ||
AK000314.1 | - | 1 | 1725 | |||
AK000317.1 | - | 1 | 675 |