Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG2 SIRNA Assays ›
Gene Symbol:
NDRG2
Gene Name:
NDRG family member 2
Gene Aliases:
SYLD
Chromosome Location:
Chr.14: 21016763 - 21070872 on Build GRCh38
UniGene ID:
Hs.525205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24168
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282211.1
NP_001269140.1
4
284
NM_001282212.1
NP_001269141.1
5
558
NM_001282213.1
NP_001269142.1
5
563
NM_001282214.1
NP_001269143.1
5
551
NM_001282215.1
NP_001269144.1
5
381
NM_001282216.1
NP_001269145.1
4
299
NM_001320329.1
NP_001307258.1
5
1216
NM_016250.2
NP_057334.1
5
406
NM_201535.1
NP_963293.1
6
448
NM_201536.1
NP_963294.1
4
339
NM_201537.1
NP_963831.1
5
381
NM_201538.1
NP_963832.1
4
316
NM_201539.1
NP_963833.1
5
358
NM_201540.1
NP_963834.1
6
425
NM_201541.1
NP_963835.1
5
383
XM_011536996.2
XP_011535298.1
6
435
XM_011536997.1
XP_011535299.1
6
436
XM_011536998.1
XP_011535300.1
6
426
XM_011536999.1
XP_011535301.1
4
831
XM_011537001.1
XP_011535303.1
5
389
XM_011537002.1
XP_011535304.1
5
535
XM_017021480.1
XP_016876969.1
6
387
XM_017021481.1
XP_016876970.1
6
426
XM_017021482.1
XP_016876971.1
6
435
XM_017021483.1
XP_016876972.1
5
319
XM_017021484.1
XP_016876973.1
5
361
XM_017021485.1
XP_016876974.1
6
426
XM_017021486.1
XP_016876975.1
6
436
XM_017021487.1
XP_016876976.1
5
345
XM_017021488.1
XP_016876977.1
5
386
XM_017021489.1
XP_016876978.1
5
491
XM_017021490.1
XP_016876979.1
4
283
XM_017021491.1
XP_016876980.1
4
318
XM_017021492.1
XP_016876981.1
5
389
XM_017021493.1
XP_016876982.1
5
535
XM_017021494.1
XP_016876983.1
6
387
XM_017021495.1
XP_016876984.1
5
319
XM_017021496.1
XP_016876985.1
4
318
GenBank mRNA
AB033074.1
-
4
268
AF087872.1
-
4
322
AF159092.3
-
5
329
AF304051.1
-
6
448
AK024521.1
-
6
381
AK057843.1
-
5
352
AK057847.1
-
4
316
AK090657.1
-
4
255
AK096999.1
-
5
1216
AK130029.1
-
4
254
AK293514.1
-
4
284
AL136574.1
-
4
293
BC010458.1
-
4
287
4215141
BC011240.1
-
5
341
4153602
BC093038.1
-
5
382
4811718
BG818977.1
-
5
362
BI759006.1
-
6
367
BM810623.1
-
5
322
BX247987.1
-
5
417
BX248031.1
-
3
245
BX647091.1
-
2
481
BX647748.1
-
5
498
CB153966.1
-
4
382
CR749252.1
-
5
551
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline