Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 40 | 5092 | ||
XM_005248729.4 | XP_005248786.1 | 40 | 5089 | |||
XM_011535918.2 | XP_011534220.1 | 37 | 4748 | |||
XM_017010989.1 | XP_016866478.1 | 42 | 5234 | |||
XM_017010990.1 | XP_016866479.1 | 39 | 4832 | |||
GenBank mRNA | AF310250.1 | - | 17 | 2099 | ||
AJ303102.1 | - | 40 | 4879 | |||
AK000712.1 | - | 13 | 1722 | |||
AK075124.1 | - | 9 | 1179 | |||
AL161957.1 | - | 17 | 4947 | |||
BC008909.2 | - | 15 | 1820 | |||
BC081569.1 | - | 18 | 2135 | |||
BC137488.1 | - | 40 | 4972 | 9021488 | ||
DQ924532.1 | - | 40 | 4866 |