Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018013.3 | NP_060483.3 | 6 | 2912 | ||
XM_005267041.4 | XP_005267098.1 | 7 | 3073 | |||
XM_005267042.4 | XP_005267099.1 | 6 | 2978 | |||
XM_011535920.2 | XP_011534222.1 | 7 | 3073 | |||
XM_011535921.2 | XP_011534223.1 | 7 | 2597 | |||
XM_011535923.2 | XP_011534225.1 | 2 | 1811 | |||
XM_017010991.1 | XP_016866480.1 | 5 | 3435 | |||
XM_017010992.1 | XP_016866481.1 | 5 | 2204 | |||
XM_017010993.1 | XP_016866482.1 | 5 | 2234 | |||
XM_017010994.1 | XP_016866483.1 | 5 | 2193 | |||
GenBank mRNA | BC113966.1 | - | 2 | 205 | ||
DQ507800.1 | - | 6 | 2408 |