Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289067.1 | NP_001275996.1 | 15 | 1827 | ||
NM_001289068.1 | NP_001275997.1 | 14 | 1644 | |||
NM_001289069.1 | NP_001275998.1 | 14 | 1593 | |||
NM_001289070.1 | NP_001275999.1 | 12 | 1395 | |||
NM_001289071.1 | NP_001276000.1 | 15 | 1733 | |||
NM_001289072.1 | NP_001276001.1 | 12 | 1299 | |||
NM_001289073.1 | NP_001276002.1 | 13 | 1624 | |||
NM_001289074.1 | NP_001276003.1 | 15 | 1608 | |||
NM_001289075.1 | NP_001276004.1 | 13 | 1492 | |||
NM_018063.4 | NP_060533.2 | 14 | 1689 | |||
GenBank mRNA | AB074174.1 | - | 2 | 137 | ||
AB102716.1 | - | 15 | 1578 | |||
AB102717.1 | - | 13 | 1469 | |||
AB102718.1 | - | 14 | 1438 | |||
AB102719.1 | - | 12 | 1144 | |||
AB102720.1 | - | 13 | 1337 | |||
AB102721.1 | - | 12 | 1240 | |||
AB102722.1 | - | 15 | 1453 | |||
AB102723.1 | - | 15 | 1672 | |||
AB113248.1 | - | 14 | 1534 | |||
AB113249.1 | - | 14 | 1534 | |||
AF155827.1 | - | 14 | 1639 | |||
AK314485.1 | - | 14 | 1678 | |||
AY007108.1 | - | 2 | 146 | |||
BC029381.1 | - | 2 | 77 | |||
BC030963.1 | - | 3 | 292 | |||
BC031004.1 | - | 3 | 292 | |||
BX538033.1 | - | 14 | 1628 |