Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172828.1 | NP_001166299.1 | 2, 3 | 287 | ||
NM_001173128.1 | NP_001166599.1 | 4, 5 | 394 | |||
NM_016258.2 | NP_057342.2 | 3, 4 | 628 | |||
GenBank mRNA | AB451335.1 | - | 3, 4 | 131 | ||
AB451484.1 | - | 3, 4 | 131 | |||
AF155095.1 | - | 3, 4 | 281 | |||
AF192968.1 | - | 3, 4 | 281 | |||
AF432214.1 | - | 2, 3 | 811 | |||
AK292581.1 | - | 3, 4 | 430 | |||
AK303833.1 | - | 2, 3 | 287 | |||
BC002559.2 | - | 3, 4 | 493 | 3139250 | ||
BP320783.1 | - | 4, 5 | 395 | |||
DC425394.1 | - | 4, 5 | 394 |