Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271844.1 | NP_001258773.1 | 3 | 350 | ||
NM_001271845.1 | NP_001258774.1 | 3 | 389 | |||
NM_001271846.1 | NP_001258775.1 | 3 | 389 | |||
NM_019020.3 | NP_061893.2 | 7 | 1505 | |||
XM_005257049.2 | XP_005257106.1 | 7 | 1516 | |||
XM_005257050.3 | XP_005257107.1 | 7 | 1519 | |||
XM_005257052.3 | XP_005257109.1 | 7 | 1522 | |||
XM_006721694.3 | XP_006721757.1 | 7 | 1522 | |||
XM_006721695.3 | XP_006721758.1 | 7 | 1522 | |||
XM_011524322.2 | XP_011522624.1 | 7 | 1480 | |||
XM_011524323.2 | XP_011522625.2 | 5 | 1430 | |||
XM_011524324.2 | XP_011522626.1 | 3 | 974 | |||
XM_017024185.1 | XP_016879674.1 | 7 | 1519 | |||
XM_017024186.1 | XP_016879675.1 | 5 | 1257 | |||
XM_017024187.1 | XP_016879676.1 | 6 | 1497 | |||
XM_017024188.1 | XP_016879677.1 | 5 | 637 | |||
GenBank mRNA | AB449902.1 | - | 6 | 1379 | ||
BC001525.2 | - | 3 | 360 | |||
BC028290.1 | - | 7 | 1495 | 4480654 | ||
BC036947.1 | - | 3 | 394 | 5392579 | ||
CR936670.1 | - | 3 | 355 |