Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016262.4 | NP_057346.1 | 3 | 239 | ||
XM_011535875.2 | XP_011534177.1 | 3 | 220 | |||
XM_017010918.1 | XP_016866407.1 | 3 | 184 | |||
GenBank mRNA | AF201334.1 | - | 3 | 125 | ||
AK026242.1 | - | 3 | 204 | |||
BC025405.2 | - | 3 | 144 | 4824105 | ||
BC031101.1 | - | 3 | 192 | 5298159 | ||
DA160210.1 | - | 3 | 239 |