Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020345.3 | NP_065078.1 | 5 | 865 | ||
XM_005265076.3 | XP_005265133.1 | 5 | 752 | |||
XM_005265077.4 | XP_005265134.1 | 4 | 720 | |||
XM_005265078.3 | XP_005265135.1 | 5 | 704 | |||
GenBank mRNA | AF229839.1 | - | 3 | 463 | ||
AK027749.1 | - | 5 | 842 | |||
BC012145.1 | - | 5 | 748 | 4549097 | ||
BC066940.1 | - | 5 | 756 | 5273965 |