Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204221.1 | NP_001191150.1 | 4 | 553 | ||
NM_020394.4 | NP_065127.4 | 4 | 553 | |||
NR_037892.1 | 4 | 553 | ||||
GenBank mRNA | BC023527.2 | - | 4 | 461 | ||
BC041082.1 | - | 4 | 490 | |||
BE743952.1 | - | 3 | 292 | |||
CN394881.1 | - | 4 | 485 |
Gene Symbol: ZNF670-ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037894.1 | 4 | 549 | |||
GenBank mRNA | AK126366.1 | - | 6 | 708 | ||
AL524527.3 | - | 4 | 472 |